SY3H580D8999 Synthonix Corporation

tert-butyl 1-amino-5-azaspiro[2.3]hexane-5-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 2007925-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59868.html

Safety Information

MSDS https://synthonix.com/59868.html
Availability details for
Quantity
  • Availability
  • Add to Cart