SY3H580D899B Synthonix Corporation

tert-butyl 4-[cis-2-aminocyclohexyl]piperazine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 2177263-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59875.html

Safety Information

MSDS https://synthonix.com/59875.html
Availability details for
Quantity
  • Availability
  • Add to Cart