SY3H580D89A2 Synthonix Corporation

(S)-2-Amino-2-(naphthalen-1-yl)ethanol hydrochloride

  • MDL number MFCD12758441
  • Cas Number : 110480-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/56576.html

Safety Information

MSDS https://synthonix.com/56576.html
Availability details for
Quantity
  • Availability
  • Add to Cart