SY3H580D89B7 Synthonix Corporation

benzyl cis-7-oxo-octahydro-1H-cyclopenta[c]pyridine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 140211-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59948.html

Safety Information

MSDS https://synthonix.com/59948.html
Availability details for
Quantity
  • Availability
  • Add to Cart