SY3H580D89BD Synthonix Corporation

benzyl 3,3-difluoro-4-hydroxy-pyrrolidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1638761-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59969.html

Safety Information

MSDS https://synthonix.com/59969.html
Availability details for
Quantity
  • Availability
  • Add to Cart