SY3H580D89C7 Synthonix Corporation

tert-butyl (3S)-3-hydroxy-2,2-dimethyl-azetidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 2227198-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59998.html

Safety Information

MSDS https://synthonix.com/59998.html
Availability details for
Quantity
  • Availability
  • Add to Cart