SY3H580D89E7 Synthonix Corporation

exo-7-(boc-amino)-3-oxa-9-aza-bicyclo[3.3.1]nonane

  • MDL number No MDL found
  • Cas Number : 1638683-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60058.html

Safety Information

MSDS https://synthonix.com/60058.html
Availability details for
Quantity
  • Availability
  • Add to Cart