SY3H580D89E9 Synthonix Corporation

1-[(tert-butoxy)carbonyl]-3,3-difluoropyrrolidine-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1822567-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60060.html

Safety Information

MSDS https://synthonix.com/60060.html
Availability details for
Quantity
  • Availability
  • Add to Cart