SY3H580D89EE Synthonix Corporation

5-[trans-2-{[(tert-butoxy)carbonyl]amino}cyclopropyl]thiophene-2-carboxylic acid

  • MDL number MFCD30723471
  • Cas Number : 2092435-93-1
  • Molecular Weight : 283.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/60067.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/60067.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart