SY3H580D89FC Synthonix Corporation

tert-butyl N-{2-azaspiro[3.3]heptan-5-yl}carbamate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1801767-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60093.html

Safety Information

MSDS https://synthonix.com/60093.html
Availability details for
Quantity
  • Availability
  • Add to Cart