SY3H580D8A0A Synthonix Corporation

tert-butyl 1-amino-3-azabicyclo[3.2.1]octane-3-carboxylate

  • MDL number MFCD31926445
  • Cas Number : 1780781-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/60129.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/60129.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart