SY3H580D8A0C Synthonix Corporation

tert-butyl 3-(6-amino-3-pyridyl)azetidine-1-carboxylate

  • MDL number MFCD24467684
  • Cas Number : 1346673-86-6
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/60137.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/60137.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart