SY3H580D8A30 Synthonix Corporation

2-cyclopropyl-5H-pyrrolo[2,3-b]pyrazine-7-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1350713-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59947.html

Safety Information

MSDS https://synthonix.com/59947.html
Availability details for
Quantity
  • Availability
  • Add to Cart