SY3H580D8A32 Synthonix Corporation

3-cyanobicyclo[1.1.1]pentane-1-carboxylic acid

  • MDL number MFCD19229172
  • Cas Number : 83249-02-9
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/60014.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/60014.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart