SY3H580D8A3C Synthonix Corporation

6-[(cyclopropylamino)methyl]-1H-pyrimidine-2,4-dione

  • MDL number No MDL found
  • Cas Number : 1339206-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60149.html

Safety Information

MSDS https://synthonix.com/60149.html
Availability details for
Quantity
  • Availability
  • Add to Cart