SY3H580D8A46 Synthonix Corporation

4-(Aminomethyl)-1-methyl-piperidin-2-one

  • MDL number MFCD17015880
  • Cas Number : 1234616-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/0719.html

Safety Information

MSDS https://synthonix.com/0719.html
Availability details for
Quantity
  • Availability
  • Add to Cart