SY3H580D8A62 Synthonix Corporation

(2,2-difluorocyclobutyl)methanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 2055839-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59993.html

Safety Information

MSDS https://synthonix.com/59993.html
Availability details for
Quantity
  • Availability
  • Add to Cart