SY3H580D8A80 Synthonix Corporation

ethyl 6-oxospiro[2.5]octane-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1447942-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60092.html

Safety Information

MSDS https://synthonix.com/60092.html
Availability details for
Quantity
  • Availability
  • Add to Cart