SY3H580D8A9C Synthonix Corporation

(4-fluoro-1-bicyclo[2.2.2]octanyl)methanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 943845-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60121.html

Safety Information

MSDS https://synthonix.com/60121.html
Availability details for
Quantity
  • Availability
  • Add to Cart