SY3H580D8A9D Synthonix Corporation

8-fluoro-5-hydroxy-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number No MDL found
  • Cas Number : 1388025-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60138.html

Safety Information

MSDS https://synthonix.com/60138.html
Availability details for
Quantity
  • Availability
  • Add to Cart