SY3H580D8A9F Synthonix Corporation

(3S)-8-Fluoro-1-methyl-3-[2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2,4(1H,3H)-quinazolinedione

  • MDL number MFCD30802489
  • Cas Number : 1643446-65-4
  • Molecular Weight : 410.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/60152.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/60152.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart