SY3H580D8AC2 Synthonix Corporation

1-methyl-1H-pyrazolo[3,4-c]pyridine-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1546503-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59713.html

Safety Information

MSDS https://synthonix.com/59713.html
Availability details for
Quantity
  • Availability
  • Add to Cart