SY3H580D8AC9 Synthonix Corporation

methyl 3-hydroxybicyclo[1.1.1]pentane-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 2092825-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59769.html

Safety Information

MSDS https://synthonix.com/59769.html
Availability details for
Quantity
  • Availability
  • Add to Cart