SY3H580D8ACF Synthonix Corporation

(3S,6R)-rel-6-methylpiperidin-3-ol hydrochloride

  • MDL number No MDL found
  • Cas Number : 67459-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59818.html

Safety Information

MSDS https://synthonix.com/59818.html
Availability details for
Quantity
  • Availability
  • Add to Cart