SY3H580D8ADC Synthonix Corporation

methyl 6-tert-butylthieno[3,2-d]pyrimidine-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 2231673-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59888.html

Safety Information

MSDS https://synthonix.com/59888.html
Availability details for
Quantity
  • Availability
  • Add to Cart