SY3H580D8AF1 Synthonix Corporation

3-methoxy-1,5-naphthyridine-4-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 1056877-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60041.html

Safety Information

MSDS https://synthonix.com/60041.html
Availability details for
Quantity
  • Availability
  • Add to Cart