SY3H580D8AF3 Synthonix Corporation

cis-2-methoxycyclopentan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1431373-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60050.html

Safety Information

MSDS https://synthonix.com/60050.html
Availability details for
Quantity
  • Availability
  • Add to Cart