SY3H580D8AF4 Synthonix Corporation

(2R)-2-(2-methoxyphenyl)-2-(oxan-4-yloxy)ethan-1-ol

  • MDL number MFCD30748064
  • Cas Number : 1434652-66-0
  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/60051.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/60051.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart