SY3H580D8B08 Synthonix Corporation

methyl 4-bromobicyclo[2.2.1]heptane-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 15448-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60159.html

Safety Information

MSDS https://synthonix.com/60159.html
Availability details for
Quantity
  • Availability
  • Add to Cart