SY3H580D8B19 Synthonix Corporation

3-isopropylbicyclo[1.1.1]pentan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1886967-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60109.html

Safety Information

MSDS https://synthonix.com/60109.html
Availability details for
Quantity
  • Availability
  • Add to Cart