SY3H580D8B2C Synthonix Corporation

trans-3-(trifluoromethyl)cyclobutanol

  • MDL number No MDL found
  • Cas Number : 2091782-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60114.html

Safety Information

MSDS https://synthonix.com/60114.html
Availability details for
Quantity
  • Availability
  • Add to Cart