SY3H580D8B34 Synthonix Corporation

2-(oxetan-2-yl)propanoic acid

  • MDL number No MDL found
  • Cas Number : 1782249-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59781.html

Safety Information

MSDS https://synthonix.com/59781.html
Availability details for
Quantity
  • Availability
  • Add to Cart