SY3H580D8B38 Synthonix Corporation

cis-octahydropyrrolo[3,4-c]pyrrol-1-one hydrochloride

  • MDL number No MDL found
  • Cas Number : 2206134-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59902.html

Safety Information

MSDS https://synthonix.com/59902.html
Availability details for
Quantity
  • Availability
  • Add to Cart