SY3H5815F3B8 Synthonix Corporation

tert-Butyl 4-hydroxy-2,2-dimethylpiperidine-1-carboxylate

  • Cas Number : 1502162-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20119.html

Safety Information

MSDS https://synthonix.com/20119.html
Availability details for
Quantity
  • Availability
  • Add to Cart