SY3H5815F3C1 Synthonix Corporation

4-Amino-2-(hydroxymethyl)pyridine

  • Cas Number : 100114-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/15900.html

Safety Information

MSDS https://synthonix.com/15900.html
Availability details for
Quantity
  • Availability
  • Add to Cart