SY3H5815F3CC Synthonix Corporation

4-Amino-6-chloro-5-methylpyrimidine

  • Cas Number : 14394-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/19834.html

Safety Information

MSDS https://synthonix.com/19834.html
Availability details for
Quantity
  • Availability
  • Add to Cart