SY3H5815F3D1 Synthonix Corporation

5-Amino-1-(tert-butyl)-1H-pyrazole-4-carbonitrile

  • Cas Number : 158001-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20487.html

Safety Information

MSDS https://synthonix.com/20487.html
Availability details for
Quantity
  • Availability
  • Add to Cart