SY3H5815F3DE Synthonix Corporation

4-Amino-5-chloro-2-methoxy-N-(piperidin-4-ylmethyl)benzamide

  • Cas Number : 220032-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22401.html

Safety Information

MSDS https://synthonix.com/22401.html
Availability details for
Quantity
  • Availability
  • Add to Cart