SY3H5815F3E3 Synthonix Corporation

2-Amino-6-bromophenol

  • Cas Number : 28165-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22810.html

Safety Information

MSDS https://synthonix.com/22810.html
Availability details for
Quantity
  • Availability
  • Add to Cart