SY3H5815F3E7 Synthonix Corporation

2-Aminopyrimidine-5-carboxylic acid

  • Cas Number : 3167-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23009.html

Safety Information

MSDS https://synthonix.com/23009.html
Availability details for
Quantity
  • Availability
  • Add to Cart