SY3H5815F3E8 Synthonix Corporation

3-Amino-5-(4-phenoxyphenyl)pyrazole-4-carbonitrile

  • Cas Number : 330792-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23083.html

Safety Information

MSDS https://synthonix.com/23083.html
Availability details for
Quantity
  • Availability
  • Add to Cart