SY3H5815F3EB Synthonix Corporation

3-Aminopyridine-2(1H)-thione

  • Cas Number : 38240-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/23408.html

Safety Information

MSDS https://synthonix.com/23408.html
Availability details for
Quantity
  • Availability
  • Add to Cart