SY3H5815F3F2 Synthonix Corporation

2-Amino-6-chloroisonicotinonitrile

  • Cas Number : 52413-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24071.html

Safety Information

MSDS https://synthonix.com/24071.html
Availability details for
Quantity
  • Availability
  • Add to Cart