SY3H5815F3F9 Synthonix Corporation

2-Amino-2-cyclohexylacetic acid

  • Cas Number : 5664-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24313.html

Safety Information

MSDS https://synthonix.com/24313.html
Availability details for
Quantity
  • Availability
  • Add to Cart