SY3H5815F3FA Synthonix Corporation

5-Amino-3-(trifluoromethyl)picolinonitrile

  • Cas Number : 573762-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24337.html

Safety Information

MSDS https://synthonix.com/24337.html
Availability details for
Quantity
  • Availability
  • Add to Cart