SY3H5815F416 Synthonix Corporation

6-Amino-8-bromo-1,7-naphthyridine

  • Cas Number : 5912-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4127.html

Safety Information

MSDS https://synthonix.com/4127.html
Availability details for
Quantity
  • Availability
  • Add to Cart