SY3H5815F418 Synthonix Corporation

3-Amino-2-bromo-5-chloropyridine

  • MDL number MFCD07368976
  • Cas Number : 90902-83-3
  • Molecular Weight : 207.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Yellow red
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4558.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/4558.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart