SY3H5815F41B Synthonix Corporation

5-Amino-1H-pyrazole-3-acetic acid

  • Cas Number : 174891-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5389.html

Safety Information

MSDS https://synthonix.com/5389.html
Availability details for
Quantity
  • Availability
  • Add to Cart