SY3H5815F41C Synthonix Corporation

2-Aminofuran-3-carbonitrile

  • Cas Number : 139370-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/5969.html

Safety Information

MSDS https://synthonix.com/5969.html
Availability details for
Quantity
  • Availability
  • Add to Cart