SY3H5815F430 Synthonix Corporation

1-Boc-3-Methanesulfonyloxymethyl-piperidine

  • Cas Number : 162166-99-6
  • Molecular Weight : 293.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to Red Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0754.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/0754.html
pictograms GHS07
hstatements H301, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart